Assay Details
| Gene Symbol: 3BETAHSD/D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036345.1 | NP_001031422.1 | 10 - 11 | 1607 | 107 | |
| NM_128183.4 | NP_180194.2 | 10 - 11 | 1661 | 107 | ||
| GenBank mRNA | AK176177.1 | - | 10 - 11 | 1607 | 107 | |
More Information
Set Membership: |
|

3BETAHSD/D2
Genomic Map