Assay Details
| Gene Symbol: 3BETAHSD/D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036345.1 | NP_001031422.1 | 2 - 3 | 459 | 101 | |
| NM_128183.4 | NP_180194.2 | 2 - 3 | 513 | 101 | ||
| GenBank mRNA | AK176177.1 | - | 2 - 3 | 459 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001036345.1 transcripts.
Set Membership: |
|

3BETAHSD/D2
Genomic Map