Assay Details
| Gene Symbol: AGL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128189.1 | NP_180200.1 | 1 - 2 | 230 | 71 | |
| GenBank mRNA | AY141233.1 | - | 1 - 2 | 230 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL33| Gene Symbol: AGL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128189.1 | NP_180200.1 | 1 - 2 | 230 | 71 | |
| GenBank mRNA | AY141233.1 | - | 1 - 2 | 230 | 71 | |
Set Membership: |
|