Assay Details
| Gene Symbol: RCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084493.1 | NP_001077962.1 | 5 - 6 | 662 | 83 | |
| NM_001084494.1 | NP_001077963.1 | 4 - 5 | 753 | 83 | ||
| NM_001336070.1 | NP_001324176.1 | 5 - 6 | 669 | 83 | ||
| NM_128200.5 | NP_565622.1 | 5 - 6 | 692 | 83 | ||
| GenBank mRNA | AK226505.1 | - | 5 - 6 | 635 | 83 | |
| AK317155.1 | - | 4 - 5 | 723 | 83 | ||
| AY086218.1 | - | 5 - 6 | 663 | 83 | ||
| AY150419.1 | - | 4 - 5 | 545 | 83 | ||
More Information
Set Membership: |
|

RCY1
Genomic Map