Assay Details
| Gene Symbol: SMU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128203.2 | NP_180214.1 | 2 - 3 | 343 | 95 | |
| GenBank mRNA | BT011249.1 | - | 2 - 3 | 239 | 95 | |
| BT012549.1 | - | 2 - 3 | 186 | 95 | ||
More Information
Set Membership: |
|

SMU2
Genomic Map