Assay Details
| Gene Symbol: PDE135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202680.1 | NP_001189609.1 | 7 - 8 | 1067 | 116 | |
| NM_001336072.1 | NP_001325095.1 | 7 - 8 | 1136 | 116 | ||
| NM_128208.5 | NP_180219.1 | 7 - 8 | 1136 | 116 | ||
| GenBank mRNA | AF370518.1 | - | 7 - 8 | 1033 | 116 | |
| BT008902.1 | - | 7 - 8 | 860 | 116 | ||
More Information
Set Membership: |
|

PDE135
Genomic Map