Assay Details
| Gene Symbol: AR781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036346.1 | NP_001031423.1 | 2 | 1246 | 160 | |
| NM_128210.4 | NP_180221.1 | 1 | 1388 | 160 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AR781| Gene Symbol: AR781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036346.1 | NP_001031423.1 | 2 | 1246 | 160 | |
| NM_128210.4 | NP_180221.1 | 1 | 1388 | 160 | ||
Set Membership: |
|