Assay Details
| Gene Symbol: PLA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128213.4 | NP_180224.1 | 1 - 2 | 346 | 98 | |
| GenBank mRNA | AY062644.1 | - | 1 - 2 | 231 | 98 | |
| AY093305.1 | - | 1 - 2 | 136 | 98 | ||
More Information
Set Membership: |
|

PLA2A
Genomic Map