Assay Details
Gene Symbol: PLA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128213.4 | NP_180224.1 | 3 - 4 | 882 | 78 | |
GenBank mRNA | AY062644.1 | - | 3 - 4 | 767 | 78 | |
AY093305.1 | - | 3 - 4 | 672 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
