Assay Details
| Gene Symbol: PLA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128213.4 | NP_180224.1 | 5 - 6 | 1348 | 105 | |
| GenBank mRNA | AY062644.1 | - | 5 - 6 | 1233 | 105 | |
| AY093305.1 | - | 5 - 6 | 1138 | 105 | ||
More Information
Set Membership: |
|

PLA2A
Genomic Map