Assay Details
| Gene Symbol: FUS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128257.4 | NP_565632.1 | 5 - 6 | 696 | 115 | |
| GenBank mRNA | AF395058.1 | - | 4 - 5 | 493 | 115 | |
| AY087622.1 | - | 5 - 6 | 581 | 115 | ||
More Information
Set Membership: |
|

FUS12
Genomic Map