Assay Details
| Gene Symbol: PAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336097.1 | NP_001323455.1 | 1 - 2 | 418 | 63 | |
| NM_128260.5 | NP_180270.1 | 1 - 2 | 264 | 63 | ||
| GenBank mRNA | AF043528.1 | - | 1 - 2 | 141 | 63 | |
| AF375455.1 | - | 1 - 2 | 131 | 63 | ||
| AY088609.1 | - | 1 - 2 | 219 | 63 | ||
| AY124806.1 | - | 1 - 2 | 98 | 63 | ||
More Information
Set Membership: |
|

PAG1
Genomic Map