Assay Details
| Gene Symbol: PAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336097.1 | NP_001323455.1 | 4 - 5 | 629 | 69 | |
| NM_128260.5 | NP_180270.1 | 4 - 5 | 475 | 69 | ||
| GenBank mRNA | AF043528.1 | - | 4 - 5 | 352 | 69 | |
| AF375455.1 | - | 4 - 5 | 342 | 69 | ||
| AY088609.1 | - | 4 - 5 | 430 | 69 | ||
| AY124806.1 | - | 4 - 5 | 309 | 69 | ||
More Information
Set Membership: |
|

PAG1
Genomic Map