Assay Details
| Gene Symbol: AT2G27130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128271.4 | NP_565637.1 | 1 - 2 | 664 | 63 | |
| GenBank mRNA | AB246330.1 | - | 1 - 2 | 361 | 63 | |
| AK175830.1 | - | 1 - 2 | 373 | 63 | ||
| AK175855.1 | - | 1 - 2 | 373 | 63 | ||
| AK175860.1 | - | 1 - 2 | 419 | 63 | ||
More Information
Set Membership: |
|

AT2G27130
Genomic Map