Assay Details
| Gene Symbol: AT2G27130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128271.4 | NP_565637.1 | 2 - 3 | 714 | 108 | |
| GenBank mRNA | AB246330.1 | - | 2 - 3 | 411 | 108 | |
| AK175830.1 | - | 2 - 3 | 423 | 108 | ||
| AK175855.1 | - | 2 - 3 | 423 | 108 | ||
| AK175860.1 | - | 2 - 3 | 469 | 108 | ||
More Information
Set Membership: |
|

AT2G27130
Genomic Map