Assay Details
| Gene Symbol: TTN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084499.2 | NP_001077968.1 | 12 - 13 | 1271 | 84 | |
| NM_001336108.1 | NP_001324367.1 | 12 - 13 | 1296 | 84 | ||
| NM_001336109.1 | NP_001324368.1 | 12 - 13 | 1230 | 84 | ||
| NM_128275.5 | NP_180285.4 | 12 - 13 | 1311 | 84 | ||
| GenBank mRNA | AK221283.1 | - | 12 - 13 | 1249 | 84 | |
More Information
Set Membership: |
|

TTN7
Genomic Map