Assay Details
| Gene Symbol: BSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202685.1 | NP_001189614.1 | 15 - 16 | 2679 | 80 | |
| NM_128279.4 | NP_180289.3 | 15 - 16 | 2552 | 80 | ||
More Information
Set Membership: |
|

BSL3
Genomic Map