Assay Details
| Gene Symbol: AT2G27460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128306.4 | NP_565651.1 | 1 - 2 | 495 | 101 | |
| GenBank mRNA | AY054495.1 | - | 1 - 2 | 342 | 101 | |
| BT010339.1 | - | 1 - 2 | 327 | 101 | ||
More Information
Set Membership: |
|

AT2G27460
Genomic Map