Assay Details
| Gene Symbol: AT2G27460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128306.4 | NP_565651.1 | 6 - 7 | 1125 | 117 | |
| GenBank mRNA | AY054495.1 | - | 6 - 7 | 972 | 117 | |
| BT010339.1 | - | 6 - 7 | 957 | 117 | ||
More Information
Set Membership: |
|

AT2G27460
Genomic Map