Assay Details
| Gene Symbol: SKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128319.4 | NP_180328.1 | 4 - 5 | 1181 | 119 | |
| GenBank mRNA | AF367297.1 | - | 4 - 5 | 1029 | 119 | |
| AY091684.1 | - | 4 - 5 | 851 | 119 | ||
More Information
Set Membership: |
|

SKD1
Genomic Map