Assay Details
| Gene Symbol: SKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128319.4 | NP_180328.1 | 7 - 8 | 1510 | 86 | |
| GenBank mRNA | AF367297.1 | - | 7 - 8 | 1358 | 86 | |
| AY091684.1 | - | 7 - 8 | 1180 | 86 | ||
More Information
Set Membership: |
|

SKD1
Genomic Map