Assay Details
| Gene Symbol: CPN60A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128359.5 | NP_180367.1 | 1 - 2 | 211 | 63 | |
| GenBank mRNA | BT002441.1 | - | 1 - 2 | 136 | 63 | |
| BT008784.1 | - | 1 - 2 | 54 | 63 | ||
More Information
Set Membership: |
|

CPN60A
Genomic Map