Assay Details
| Gene Symbol: CSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128379.4 | NP_565666.1 | 6 - 7 | 706 | 65 | |
| GenBank mRNA | AK227096.1 | - | 6 - 7 | 617 | 65 | |
| AY064050.1 | - | 6 - 7 | 616 | 65 | ||
| AY133756.1 | - | 6 - 7 | 570 | 65 | ||
More Information
Set Membership: |
|

CSD2
Genomic Map