Assay Details
| Gene Symbol: AT2G28370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128397.3 | NP_565671.1 | 3 - 4 | 682 | 67 | |
| GenBank mRNA | AF375441.1 | - | 3 - 4 | 618 | 67 | |
| AY143955.1 | - | 2 - 3 | 348 | 67 | ||
More Information
Set Membership: |
|

AT2G28370
Genomic Map