Assay Details
| Gene Symbol: DRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128398.4 | NP_565672.1 | 1 - 2 | 573 | 88 | |
| GenBank mRNA | AY034976.1 | - | 1 - 2 | 498 | 88 | |
| AY062965.1 | - | 1 - 2 | 247 | 88 | ||
More Information
Set Membership: |
|

DRB2
Genomic Map