Assay Details
| Gene Symbol: AT2G28480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128408.3 | NP_180415.1 | 4 - 5 | 1090 | 119 | |
| GenBank mRNA | AK226713.1 | - | 4 - 5 | 1013 | 119 | |
| BT011753.1 | - | 3 - 4 | 802 | 119 | ||
More Information
Set Membership: |
|

AT2G28480
Genomic Map