Assay Details
| Gene Symbol: PK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036362.3 | NP_001031439.1 | 3 - 4 | 722 | 72 | |
| NM_001036363.2 | NP_001031440.1 | 3 - 4 | 753 | 72 | ||
| NM_128452.4 | NP_180459.2 | 2 - 3 | 711 | 72 | ||
| GenBank mRNA | BT029755.1 | - | 2 - 3 | 529 | 72 | |
More Information
Set Membership: |
|

PK1B
Genomic Map