Assay Details
| Gene Symbol: SAG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036365.1 | NP_001031442.1 | 2 - 3 | 303 | 115 | |
| NM_128489.3 | NP_180496.1 | 2 - 3 | 423 | 115 | ||
| NM_201829.2 | NP_973558.1 | 2 - 3 | 423 | 115 | ||
| GenBank mRNA | AY065111.1 | - | 2 - 3 | 331 | 115 | |
| AY081642.1 | - | 2 - 3 | 301 | 115 | ||
| AY085321.1 | - | 2 - 3 | 423 | 115 | ||
More Information
Set Membership: |
|

SAG13
Genomic Map