Assay Details
| Gene Symbol: TIM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084507.2 | NP_001077976.1 | 3 - 4 | 166 | 90 | |
| NM_128506.3 | NP_565682.1 | 2 - 3 | 188 | 90 | ||
| GenBank mRNA | AF150093.1 | - | 2 - 3 | 99 | 90 | |
| AK228282.1 | - | 3 - 4 | 155 | 90 | ||
| BT004739.1 | - | 2 - 3 | 99 | 90 | ||
More Information
Set Membership: |
|

TIM10
Genomic Map