Assay Details
| Gene Symbol: ABCG31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128548.4 | NP_180555.2 | 11 - 12 | 2262 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCG31| Gene Symbol: ABCG31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128548.4 | NP_180555.2 | 11 - 12 | 2262 | 82 | |
Set Membership: |
|