Assay Details
| Gene Symbol: ABCG31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128548.4 | NP_180555.2 | 19 - 20 | 4183 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCG31| Gene Symbol: ABCG31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128548.4 | NP_180555.2 | 19 - 20 | 4183 | 89 | |
Set Membership: |
|