Assay Details
| Gene Symbol: NADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338499.1 | NP_001319603.1 | 3 - 4 | 693 | 88 | |
| NM_001338500.1 | NP_001327317.1 | 3 - 4 | 607 | 88 | ||
| NM_001338501.1 | NP_001327316.1 | 3 - 4 | 580 | 88 | ||
| NM_202618.3 | NP_974347.1 | 3 - 4 | 542 | 88 | ||
| GenBank mRNA | AK176132.1 | - | 3 - 4 | 372 | 88 | |
| AY383545.1 | - | 3 - 4 | 402 | 88 | ||
More Information
Set Membership: |
|

NADK1
Genomic Map