Assay Details
| Gene Symbol: NADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338499.1 | NP_001319603.1 | 5 - 6 | 989 | 94 | |
| NM_001338500.1 | NP_001327317.1 | 5 - 6 | 903 | 94 | ||
| NM_001338501.1 | NP_001327316.1 | 5 - 6 | 876 | 94 | ||
| NM_202618.3 | NP_974347.1 | 5 - 6 | 838 | 94 | ||
| GenBank mRNA | AK176132.1 | - | 5 - 6 | 668 | 94 | |
| AY383545.1 | - | 5 - 6 | 698 | 94 | ||
More Information
Set Membership: |
|

NADK1
Genomic Map