Assay Details
| Gene Symbol: BBX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113009.3 | NP_188752.1 | 1 | 941 | 61 | |
| GenBank mRNA | AK117844.1 | - | 1 | 755 | 61 | |
| BT005244.1 | - | 1 | 604 | 61 | ||
More Information
Set Membership: |
|

BBX32
Genomic Map