Assay Details
| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 11 - 12 | 1432 | 121 | |
| GenBank mRNA | AK227228.1 | - | 11 - 12 | 1389 | 121 | |
More Information
Set Membership: |
|

ACA9
Genomic Map