Assay Details
| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 22 - 23 | 2445 | 141 | |
| GenBank mRNA | AK227228.1 | - | 22 - 23 | 2402 | 141 | |
More Information
Set Membership: |
|

ACA9
Genomic Map