Assay Details
| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 27 - 28 | 3070 | 81 | |
| GenBank mRNA | AK227228.1 | - | 27 - 28 | 3027 | 81 | |
More Information
Set Membership: |
|

ACA9
Genomic Map