Assay Details
| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 30 - 31 | 3355 | 72 | |
| GenBank mRNA | AK227228.1 | - | 30 - 31 | 3312 | 72 | |
More Information
Set Membership: |
|

ACA9
Genomic Map