Assay Details
| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 6 - 7 | 1035 | 62 | |
| GenBank mRNA | AK227228.1 | - | 6 - 7 | 992 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACA9| Gene Symbol: ACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113013.5 | NP_188755.2 | 6 - 7 | 1035 | 62 | |
| GenBank mRNA | AK227228.1 | - | 6 - 7 | 992 | 62 | |
Set Membership: |
|