Assay Details
| Gene Symbol: MED6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161161.1 | NP_001154633.1 | 2 - 3 | 470 | 78 | |
| NM_113030.5 | NP_188772.2 | 2 - 3 | 470 | 78 | ||
| GenBank mRNA | AY070104.1 | - | 2 - 3 | 468 | 78 | |
| AY091238.1 | - | 2 - 3 | 434 | 78 | ||
More Information
Set Membership: |
|

MED6
Genomic Map