Assay Details
| Gene Symbol: CERK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113058.4 | NP_566689.2 | 1 - 2 | 784 | 99 | |
| GenBank mRNA | AB367524.1 | - | 1 - 2 | 643 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERK1| Gene Symbol: CERK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113058.4 | NP_566689.2 | 1 - 2 | 784 | 99 | |
| GenBank mRNA | AB367524.1 | - | 1 - 2 | 643 | 99 | |
Set Membership: |
|