Assay Details
| Gene Symbol: CERK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113058.4 | NP_566689.2 | 7 - 8 | 1357 | 158 | |
| GenBank mRNA | AB367524.1 | - | 7 - 8 | 1216 | 158 | |
More Information
Set Membership: |
|

CERK1
Genomic Map