Assay Details
| Gene Symbol: CERK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113058.4 | NP_566689.2 | 8 - 9 | 1513 | 119 | |
| GenBank mRNA | AB367524.1 | - | 8 - 9 | 1372 | 119 | |
More Information
Set Membership: |
|

CERK1
Genomic Map