Assay Details
| Gene Symbol: ATXR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113077.5 | NP_188819.2 | 4 - 5 | 793 | 69 | |
| GenBank mRNA | AK221711.1 | - | 4 - 5 | 730 | 69 | |
| BT020260.1 | - | 4 - 5 | 677 | 69 | ||
| BT021112.1 | - | 4 - 5 | 670 | 69 | ||
More Information
Set Membership: |
|

ATXR2
Genomic Map