Assay Details
| Gene Symbol: BBX31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113085.4 | NP_188827.1 | 1 | 556 | 76 | |
| GenBank mRNA | AB493627.1 | - | 1 | 286 | 76 | |
| AY086564.1 | - | 1 | 356 | 76 | ||
More Information
Set Membership: |
|

BBX31
Genomic Map