Assay Details
| Gene Symbol: FHY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125201.2 | NP_001118673.1 | 6 - 7 | 2875 | 64 | |
| NM_113114.3 | NP_188856.2 | 6 - 7 | 2529 | 64 | ||
More Information
Set Membership: |
|

FHY3
Genomic Map