Assay Details
| Gene Symbol: AT3G22270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113125.4 | NP_188866.1 | 1 - 2 | 380 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G22270| Gene Symbol: AT3G22270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113125.4 | NP_188866.1 | 1 - 2 | 380 | 84 | |
Set Membership: |
|