Assay Details
| Gene Symbol: OASA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338588.1 | NP_001326025.1 | 3 - 4 | 207 | 67 | |
| NM_113145.5 | NP_188885.4 | 1 - 2 | 119 | 67 | ||
| GenBank mRNA | BT031364.1 | - | 3 - 4 | 207 | 67 | |
More Information
Set Membership: |
|

OASA2
Genomic Map