Assay Details
| Gene Symbol: LFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113199.5 | NP_566721.1 | 1 - 2 | 270 | 84 | |
| GenBank mRNA | AY072191.1 | - | 1 - 2 | 221 | 84 | |
| AY096422.1 | - | 1 - 2 | 196 | 84 | ||
More Information
Set Membership: |
|

LFR
Genomic Map