Assay Details
| Gene Symbol: LFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113199.5 | NP_566721.1 | 3 - 4 | 576 | 79 | |
| GenBank mRNA | AY072191.1 | - | 3 - 4 | 527 | 79 | |
| AY096422.1 | - | 3 - 4 | 502 | 79 | ||
More Information
Set Membership: |
|

LFR
Genomic Map